4-(3-Piperidinylmethyl)-2-piperazinone dihydrochloride

Supplier: Matrix Scientific

051425-500MG 051425-2.5G
101895-592EA 364.62 USD
101895-592 101895-594
4-(3-Piperidinylmethyl)-2-piperazinone dihydrochloride
4-(3-Piperidinylmethyl)-2-piperazinone dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁Cl₂N₃O
MW: 270.2 g/mol
MDL Number: MFCD13561497
CAS Number: 1220019-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR