1-(3-Piperidinylmethyl)azepane dihydrochloride
Supplier: Matrix Scientific
051424-500MG
051424-2.5G
101895-588EA
364.62
USD
101895-588
101895-590
1-(3-Piperidinylmethyl)azepane dihydrochloride
1-(3-Piperidinylmethyl)azepane dihydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₆Cl₂N₂ |
MDL Number:
MFCD13469303 CAS Number: 1211495-72-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...