1-Benzyl-4-(3-piperidinylmethyl)piperazine dihydrochloride

Supplier: Matrix Scientific

051423-500MG 051423-2.5G
101895-584EA 364.62 USD
101895-584 101895-586
1-Benzyl-4-(3-piperidinylmethyl)piperazine dihydrochloride
1-Benzyl-4-(3-piperidinylmethyl)piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₇H₂₉Cl₂N₃
MDL Number: MFCD13561496
CAS Number: 1220029-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR