1-Methyl-4-(3-piperidinylmethyl)piperazine dihydrochloride

Supplier: Matrix Scientific

051422-500MG 051422-2.5G
101895-580EA 364.62 USD
101895-580 101895-582
1-Methyl-4-(3-piperidinylmethyl)piperazine dihydrochloride
1-Methyl-4-(3-piperidinylmethyl)piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₅Cl₂N₃
MDL Number: MFCD13561495
CAS Number: 1220029-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR