4-Methyl-1-(3-piperidinylmethyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051421-500MG 051421-2.5G
101895-576EA 364.62 USD
101895-576 101895-578
4-Methyl-1-(3-piperidinylmethyl)piperidine dihydrochloride
4-Methyl-1-(3-piperidinylmethyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆Cl₂N₂
MW: 269.26 g/mol
MDL Number: MFCD13561494
CAS Number: 1211450-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR