2-Methyl-1-(3-piperidinylmethyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051419-500MG 051419-2.5G
101895-568EA 364.62 USD
101895-568 101895-570
2-Methyl-1-(3-piperidinylmethyl)piperidine dihydrochloride
2-Methyl-1-(3-piperidinylmethyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆Cl₂N₂
MDL Number: MFCD13561492
CAS Number: 1211474-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR