2-[Ethyl(3-piperidinylmethyl)amino]-1-ethanol dihydrochloride

Supplier: Matrix Scientific

051417-500MG 051417-2.5G
101895-560EA 364.62 USD
101895-560 101895-562
2-[Ethyl(3-piperidinylmethyl)amino]-1-ethanol dihydrochloride
2-[Ethyl(3-piperidinylmethyl)amino]-1-ethanol dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₄Cl₂N₂O
MW: 259.22 g/mol
MDL Number: MFCD13561490
CAS Number: 1220019-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR