N-Methyl-N-(3-piperidinylmethyl)-1-butanamine dihydrochloride

Supplier: Matrix Scientific

051416-500MG 051416-2.5G
101895-556EA 364.62 USD
101895-556 101895-558
N-Methyl-N-(3-piperidinylmethyl)-1-butanamine dihydrochloride
N-Methyl-N-(3-piperidinylmethyl)-1-butanamine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₆Cl₂N₂
MW: 257.25 g/mol
MDL Number: MFCD13561489
CAS Number: 1219979-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR