4-(3-Piperidinylmethyl)morpholine dihydrochloride

Supplier: Matrix Scientific

051414-500MG 051414-2.5G
101895-548EA 364.62 USD
101895-548 101895-550
4-(3-Piperidinylmethyl)morpholine dihydrochloride
4-(3-Piperidinylmethyl)morpholine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂Cl₂N₂O
MDL Number: MFCD13250093
CAS Number: 81310-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR