1-(3-Piperidinylmethyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051413-500MG 051413-2.5G
101895-544EA 364.62 USD
101895-544 101895-546
1-(3-Piperidinylmethyl)piperidine dihydrochloride
1-(3-Piperidinylmethyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂
MDL Number: MFCD13561487
CAS Number: 81310-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR