N-Ethyl-N-(3-piperidinylmethyl)-1-ethanamine dihydrochloride

Supplier: Matrix Scientific

051411-500MG 051411-2.5G
101895-536EA 364.62 USD
101895-536 101895-538
N-Ethyl-N-(3-piperidinylmethyl)-1-ethanamine dihydrochloride
N-Ethyl-N-(3-piperidinylmethyl)-1-ethanamine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₄Cl₂N₂
MW: 243.22 g/mol
MDL Number: MFCD13561485
CAS Number: 1220021-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR