3-(1-Pyrrolidinylmethyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051410-500MG 051410-2.5G
101895-532EA 364.62 USD
101895-532 101895-534
3-(1-Pyrrolidinylmethyl)piperidine dihydrochloride
3-(1-Pyrrolidinylmethyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂Cl₂N₂
MDL Number: MFCD13250091
CAS Number: 514842-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR