N,N-Dimethyl(3-piperidinyl)methanamine dihydrochloride
Supplier: Matrix Scientific
Synonyms:
N,N-Dimethyl-1-piperidin-3-ylmethanamine
051409-500MG
051409-2.5G
101895-528EA
364.62
USD
101895-528
101895-530
N,N-Dimethyl(3-piperidinyl)methanamine dihydrochloride
N,N-Dimethyl(3-piperidinyl)methanamine dihydrochloride
Hazard: Irritant
Formula:
C₈H₁₈N₂·2HCl MW: 215.17 g/mol |
MDL Number:
MFCD13561484 CAS Number: 90203-05-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...