N-1-Benzyl-4-bromo-N-1-ethyl-1,2-benzenediamine

Supplier: Matrix Scientific

051407-500MG 051407-2.5G
101895-520EA 364.62 USD
101895-520 101895-522
N-1-Benzyl-4-bromo-N-1-ethyl-1,2-benzenediamine
N-1-Benzyl-4-bromo-N-1-ethyl-1,2-benzenediamine

Hazard: Irritant

Formula: C₁₅H₁₇BrN₂
MDL Number: MFCD11195374
CAS Number: 1039958-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR