5-Bromo-2-[3,4-dihydro-1(2H)-quinolinyl]aniline

Supplier: Matrix Scientific

051406-500MG 051406-2.5G
101895-516EA 364.62 USD
101895-516 101895-518
5-Bromo-2-[3,4-dihydro-1(2H)-quinolinyl]aniline
5-Bromo-2-[3,4-dihydro-1(2H)-quinolinyl]aniline

Hazard: Irritant

Formula: C₁₅H₁₅BrN₂
MDL Number: MFCD11127761
CAS Number: 112631-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR