5-Bromo-2-(2,3-dihydro-1H-indol-1-yl)phenylamine

Supplier: Matrix Scientific

051405-500MG 051405-2.5G
101895-512EA 364.62 USD
101895-512 101895-514
5-Bromo-2-(2,3-dihydro-1H-indol-1-yl)phenylamine
5-Bromo-2-(2,3-dihydro-1H-indol-1-yl)phenylamine

Hazard: Irritant

Formula: C₁₄H₁₃BrN₂
MDL Number: MFCD09949421
CAS Number: 1016789-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR