2-[4-(2-Amino-4-bromophenyl)-1-piperazinyl]-1-ethanol
Supplier: Matrix Scientific
051403-500MG
051403-2.5G
101895-504EA
364.62
USD
101895-504
101895-506
2-[4-(2-Amino-4-bromophenyl)-1-piperazinyl]-1-ethanol
2-[4-(2-Amino-4-bromophenyl)-1-piperazinyl]-1-ethanol
Hazard: Irritant
Formula:
C₁₂H₁₈BrN₃O |
MDL Number:
MFCD12421865 CAS Number: 1155621-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...