N-(2-Amino-4-bromophenyl)-N-cyclohexyl-N-ethylamine

Supplier: Matrix Scientific

051402-500MG 051402-2.5G
101895-500EA 364.62 USD
101895-500 101895-502
N-(2-Amino-4-bromophenyl)-N-cyclohexyl-N-ethylamine
N-(2-Amino-4-bromophenyl)-N-cyclohexyl-N-ethylamine

Hazard: Irritant

Formula: C₁₄H₂₁BrN₂
MDL Number: MFCD11127763
CAS Number: 1019354-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR