N-(2-Amino-4-bromophenyl)-N-cyclohexyl-N-methylamine

Supplier: Matrix Scientific

051399-500MG 051399-2.5G
101895-488EA 364.62 USD
101895-488 101895-490
N-(2-Amino-4-bromophenyl)-N-cyclohexyl-N-methylamine
N-(2-Amino-4-bromophenyl)-N-cyclohexyl-N-methylamine

Hazard: Irritant

Formula: C₁₃H₁₉BrN₂
MDL Number: MFCD09940568
CAS Number: 1016813-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR