4-Bromo-N-1,N-1-dipropyl-1,2-benzenediamine

Supplier: Matrix Scientific

051398-500MG 051398-2.5G
101895-484EA 364.62 USD
101895-484 101895-486
4-Bromo-N-1,N-1-dipropyl-1,2-benzenediamine
4-Bromo-N-1,N-1-dipropyl-1,2-benzenediamine

Hazard: Irritant

Formula: C₁₂H₁₉BrN₂
MDL Number: MFCD09948934
CAS Number: 1016805-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR