2-(4-Benzyl-1-piperazinyl)-5-bromophenylamine

Supplier: Matrix Scientific

051394-500MG 051394-2.5G
101895-468EA 364.62 USD
101895-468 101895-470
2-(4-Benzyl-1-piperazinyl)-5-bromophenylamine
2-(4-Benzyl-1-piperazinyl)-5-bromophenylamine

Hazard: Irritant

Formula: C₁₇H₂₀BrN₃
MDL Number: MFCD09947954
CAS Number: 883908-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR