5-Bromo-2-(4-methyl-1-piperazinyl)aniline
Supplier: Matrix Scientific
051393-500MG
051393-2.5G
101895-464EA
364.62
USD
101895-464
101895-466
5-Bromo-2-(4-methyl-1-piperazinyl)aniline
5-Bromo-2-(4-methyl-1-piperazinyl)aniline
Hazard: Irritant
Formula:
C₁₁H₁₆BrN₃ |
MDL Number:
MFCD06660761 CAS Number: 885461-06-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...