5-Bromo-2-(2-methyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051390-500MG 051390-2.5G
101895-452EA 364.62 USD
101895-452 101895-454
5-Bromo-2-(2-methyl-1-piperidinyl)aniline
5-Bromo-2-(2-methyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₇BrN₂
MDL Number: MFCD09935983
CAS Number: 1016849-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR