N-1,N-1-Diallyl-4-bromo-1,2-benzenediamine
Supplier: Matrix Scientific
051389-500MG
051389-2.5G
101895-448EA
364.62
USD
101895-448
101895-450
N-1,N-1-Diallyl-4-bromo-1,2-benzenediamine
N-1,N-1-Diallyl-4-bromo-1,2-benzenediamine
Hazard: Irritant
Formula:
C₁₂H₁₅BrN₂ |
MDL Number:
MFCD13561482 CAS Number: 1220016-76-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...