1-(2-Amino-4-bromophenyl)-3-pyrrolidinol

Supplier: Matrix Scientific

051386-500MG 051386-2.5G
101895-436EA 364.62 USD
101895-436 101895-438
1-(2-Amino-4-bromophenyl)-3-pyrrolidinol
1-(2-Amino-4-bromophenyl)-3-pyrrolidinol

Hazard: Irritant

Formula: C₁₀H₁₃BrN₂O
MDL Number: MFCD13561481
CAS Number: 1219957-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR