5-Bromo-2-(4-morpholinyl)aniline
Supplier: Matrix Scientific
051385-500MG
051385-2.5G
101895-432EA
364.62
USD
101895-432
101895-434
5-Bromo-2-(4-morpholinyl)aniline
5-Bromo-2-(4-morpholinyl)aniline
Hazard: Irritant
Formula:
C₁₀H₁₃BrN₂O MW: 257.14 g/mol |
MDL Number:
MFCD06660760 CAS Number: 91062-48-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...