5-Bromo-2-(1-piperidinyl)aniline

Supplier: Matrix Scientific

051384-500MG 051384-2.5G
101895-428EA 364.62 USD
101895-428 101895-430
5-Bromo-2-(1-piperidinyl)aniline
5-Bromo-2-(1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₁H₁₅BrN₂
MDL Number: MFCD09951284
CAS Number: 1016877-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR