5-Bromo-2-(1-pyrrolidinyl)aniline

Supplier: Matrix Scientific

051381-500MG 051381-2.5G
101895-416EA 364.62 USD
101895-416 101895-418
5-Bromo-2-(1-pyrrolidinyl)aniline
5-Bromo-2-(1-pyrrolidinyl)aniline

Hazard: Irritant

Formula: C₁₀H₁₃BrN₂
MDL Number: MFCD09939470
CAS Number: 1016795-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR