4-Bromo-N-1,N-1-dimethyl-1,2-benzenediamine

Supplier: Matrix Scientific

051380-500MG 051380-2.5G
101895-412EA 364.62 USD
101895-412 101895-414
4-Bromo-N-1,N-1-dimethyl-1,2-benzenediamine
4-Bromo-N-1,N-1-dimethyl-1,2-benzenediamine

Hazard: Irritant

Formula: C₈H₁₁BrN₂
MDL Number: MFCD06660759
CAS Number: 183251-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR