4-[3,4-Dihydro-1(2H)-quinolinyl]-2-methylaniline
Supplier: Matrix Scientific
051377-500MG
051377-2.5G
101895-400EA
364.62
USD
101895-400
101895-402
4-[3,4-Dihydro-1(2H)-quinolinyl]-2-methylaniline
4-[3,4-Dihydro-1(2H)-quinolinyl]-2-methylaniline
Hazard: Irritant
Formula:
C₁₆H₁₈N₂ |
MDL Number:
MFCD11979471 CAS Number: 1154288-79-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...