4-[3,4-Dihydro-2(1H)-isoquinolinyl]-2-methylaniline

Supplier: Matrix Scientific

051375-500MG 051375-2.5G
101895-392EA 364.62 USD
101895-392 101895-394
4-[3,4-Dihydro-2(1H)-isoquinolinyl]-2-methylaniline
4-[3,4-Dihydro-2(1H)-isoquinolinyl]-2-methylaniline

Hazard: Irritant

Formula: C₁₆H₁₈N₂
MDL Number: MFCD12410389
CAS Number: 1155005-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR