2-[4-(4-Amino-3-methylphenyl)-1-piperazinyl]-1-ethanol

Supplier: Matrix Scientific

051374-500MG 051374-2.5G
101895-388EA 364.62 USD
101895-388 101895-390
2-[4-(4-Amino-3-methylphenyl)-1-piperazinyl]-1-ethanol
2-[4-(4-Amino-3-methylphenyl)-1-piperazinyl]-1-ethanol

Hazard: Irritant

Formula: C₁₃H₂₁N₃O
MDL Number: MFCD12421859
CAS Number: 1154310-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR