N-4-Cyclohexyl-N-4-ethyl-2-methyl-1,4-benzenediamine
Supplier: Matrix Scientific
051373-500MG
051373-2.5G
101895-384EA
364.62
USD
101895-384
101895-386
N-4-Cyclohexyl-N-4-ethyl-2-methyl-1,4-benzenediamine
N-4-Cyclohexyl-N-4-ethyl-2-methyl-1,4-benzenediamine
Hazard: Irritant
Formula:
C₁₅H₂₄N₂ |
MDL Number:
MFCD12410393 CAS Number: 1155985-58-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...