4-(2-Ethyl-1-piperidinyl)-2-methylaniline

Supplier: Matrix Scientific

051371-500MG 051371-2.5G
101895-376EA 364.62 USD
101895-376 101895-378
4-(2-Ethyl-1-piperidinyl)-2-methylaniline
4-(2-Ethyl-1-piperidinyl)-2-methylaniline

Hazard: Irritant

Formula: C₁₄H₂₂N₂
MDL Number: MFCD12169220
CAS Number: 1154644-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR