4-(4-Benzyl-1-piperazinyl)-2-methylphenylamine

Supplier: Matrix Scientific

051365-500MG 051365-2.5G
101895-352EA 364.62 USD
101895-352 101895-354
4-(4-Benzyl-1-piperazinyl)-2-methylphenylamine
4-(4-Benzyl-1-piperazinyl)-2-methylphenylamine

Hazard: Irritant

Formula: C₁₈H₂₃N₃
MDL Number: MFCD13561479
CAS Number: 864932-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR