2-Methyl-4-(4-methyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051363-500MG 051363-2.5G
101895-344EA 364.62 USD
101895-344 101895-346
2-Methyl-4-(4-methyl-1-piperidinyl)aniline
2-Methyl-4-(4-methyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₃H₂₀N₂
MW: 204.32 g/mol
MDL Number: MFCD12169090
CAS Number: 1155005-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR