2-Methyl-4-(2-methyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051361-500MG 051361-2.5G
101895-336EA 364.62 USD
101895-336 101895-338
2-Methyl-4-(2-methyl-1-piperidinyl)aniline
2-Methyl-4-(2-methyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₃H₂₀N₂
MDL Number: MFCD12169091
CAS Number: 1155017-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR