N-4-Butyl-(N-4,2-dimethyl)-1,4-benzenediamine

Supplier: Matrix Scientific

051358-500MG 051358-2.5G
101895-324EA 364.62 USD
101895-324 101895-326
N-4-Butyl-(N-4,2-dimethyl)-1,4-benzenediamine
N-4-Butyl-(N-4,2-dimethyl)-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₂H₂₀N₂
MDL Number: MFCD12169089
CAS Number: 1155005-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR