1-(4-Amino-3-methylphenyl)-3-pyrrolidinol

Supplier: Matrix Scientific

051357-500MG 051357-2.5G
101895-320EA 364.62 USD
101895-320 101895-322
1-(4-Amino-3-methylphenyl)-3-pyrrolidinol
1-(4-Amino-3-methylphenyl)-3-pyrrolidinol

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
MDL Number: MFCD11848952
CAS Number: 143525-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR