2-Methyl-4-(4-morpholinyl)aniline

Supplier: Matrix Scientific

Synonyms: 2-Methyl-4-(4-morpholinyl)benzenamine

051356-500MG 051356-2.5G
101895-316EA 93.85 USD
101895-316 101895-318
2-Methyl-4-(4-morpholinyl)aniline
2-Methyl-4-(4-morpholinyl)aniline

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
MW: 192.26 g/mol
MDL Number: MFCD10686817
CAS Number: 581-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR