N-4,N-4-Diethyl-2-methyl-1,4-benzenediamine

Supplier: Matrix Scientific

051353-500MG 051353-2.5G
101895-304EA 461.54 USD
101895-304 101895-306
N-4,N-4-Diethyl-2-methyl-1,4-benzenediamine
N-4,N-4-Diethyl-2-methyl-1,4-benzenediamine

Hazard: Irritant

Formula: C₁₁H₁₈N₂
MW: 178.28 g/mol
MDL Number: MFCD00137576
CAS Number: 148-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR