2-Methyl-4-(1-pyrrolidinyl)aniline

Supplier: Matrix Scientific

051352-500MG 051352-2.5G
101895-300EA 364.62 USD
101895-300 101895-302
2-Methyl-4-(1-pyrrolidinyl)aniline
2-Methyl-4-(1-pyrrolidinyl)aniline

Hazard: Irritant

Formula: C₁₁H₁₆N₂
MW: 176.26 g/mol
MDL Number: MFCD06337021
CAS Number: 143525-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR