4-[3,4-Dihydro-2(1H)-isoquinolinyl]-3-fluoroaniline

Supplier: Matrix Scientific

051346-500MG 051346-2.5G
101895-276EA 364.62 USD
101895-276 101895-278
4-[3,4-Dihydro-2(1H)-isoquinolinyl]-3-fluoroaniline
4-[3,4-Dihydro-2(1H)-isoquinolinyl]-3-fluoroaniline

Hazard: Irritant

Formula: C₁₅H₁₅FN₂
MDL Number: MFCD08699586
CAS Number: 449175-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR