2-[4-(4-Amino-2-fluorophenyl)-1-piperazinyl]-1-ethanol
Supplier: Matrix Scientific
051345-500MG
051345-2.5G
101895-272EA
364.62
USD
101895-272
101895-274
2-[4-(4-Amino-2-fluorophenyl)-1-piperazinyl]-1-ethanol
2-[4-(4-Amino-2-fluorophenyl)-1-piperazinyl]-1-ethanol
Hazard: Irritant
Formula:
C₁₂H₁₈FN₃O |
MDL Number:
MFCD12421863 CAS Number: 1155522-89-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...