1-(4-Amino-2-fluorophenyl)-4-piperidinol

Supplier: Matrix Scientific

051339-500MG 051339-2.5G
101895-248EA 335.38 USD
101895-248 101895-250
1-(4-Amino-2-fluorophenyl)-4-piperidinol
1-(4-Amino-2-fluorophenyl)-4-piperidinol

Hazard: Irritant

Formula: C₁₁H₁₅FN₂O
MDL Number: MFCD11193222
CAS Number: 873537-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR