4-(4-Amino-2-fluorophenyl)-2-piperazinone

Supplier: Matrix Scientific

051338-500MG 051338-2.5G
101895-244EA 364.62 USD
101895-244 101895-246
4-(4-Amino-2-fluorophenyl)-2-piperazinone
4-(4-Amino-2-fluorophenyl)-2-piperazinone

Hazard: Irritant

Formula: C₁₀H₁₂FN₃O
MDL Number: MFCD11193221
CAS Number: 850369-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR