3-Fluoro-4-(4-methyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051335-500MG 051335-2.5G
101895-232EA 364.62 USD
101895-232 101895-234
3-Fluoro-4-(4-methyl-1-piperidinyl)aniline
3-Fluoro-4-(4-methyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₇FN₂
MDL Number: MFCD08699559
CAS Number: 250371-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR