3-Fluoro-4-(3-methyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051334-500MG 051334-2.5G
101895-228EA 364.62 USD
101895-228 101895-230
3-Fluoro-4-(3-methyl-1-piperidinyl)aniline
3-Fluoro-4-(3-methyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₇FN₂
MDL Number: MFCD08699560
CAS Number: 937597-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR