3-Fluoro-4-(2-methyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051333-500MG 051333-2.5G
101895-224EA 364.62 USD
101895-224 101895-226
3-Fluoro-4-(2-methyl-1-piperidinyl)aniline
3-Fluoro-4-(2-methyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₇FN₂
MDL Number: MFCD10691930
CAS Number: 952906-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR