2-[4-Amino(ethyl)-2-fluoroanilino]-1-ethanol

Supplier: Matrix Scientific

051331-500MG 051331-2.5G
101895-216EA 364.62 USD
101895-216 101895-218
2-[4-Amino(ethyl)-2-fluoroanilino]-1-ethanol
2-[4-Amino(ethyl)-2-fluoroanilino]-1-ethanol

Hazard: Irritant

Formula: C₁₀H₁₅FN₂O
MDL Number: MFCD12813172
CAS Number: 1179154-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR